Anti-CD339 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: HMJ1-29]

Supplier: BioLegend
130908 130907 130907 130908
10757-160EA 165.11 USD
CA10757-162 10757-160 CA10757-160 10757-162
Anti-CD339 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: HMJ1-29]
Antibodies
PE anti-mouse CD339 (Jagged 1) [HMJ1-29]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD339
Clonality: Monoclonal
Clone: HMJ1-29
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR