Anti-CD335 Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: 29A1.4]

Supplier: BioLegend
137621 137621
10756-444EA 360.03 USD
CA10756-444 10756-444
Anti-CD335 Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: 29A1.4]
Antibodies
Brilliant Violet 711™ anti-mouse CD335 (NKp46) [29A1.4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD335
Clonality: Monoclonal
Clone: 29A1.4
Conjugation: Brilliant Violet® 711
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR