Anti-CD335 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 29A1.4]

Supplier: BioLegend
137618 137618 137617 137617
137618-BLEA 501.17 USD
137618-BL CA137617-BL 137617-BL CA137618-BL
Anti-CD335 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 29A1.4]
Antibodies
PE/Cy7 anti-mouse CD335 (NKp46) [29A1.4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD335
Clonality: Monoclonal
Clone: 29A1.4
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR