Anti-CD335 Rat Monoclonal Antibody (LEAF) [clone: 29A1.4]

Supplier: BioLegend
137614 137614 137613 137613
137614-BLEA 372.75 USD
CA137614-BL CA137613-BL 137614-BL 137613-BL
Anti-CD335 Rat Monoclonal Antibody (LEAF) [clone: 29A1.4]
LEAF™ Purified anti-mouse CD335 (NKp46) [29A1.4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC, Activ; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining, it is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD335
Clonality: Monoclonal
Clone: 29A1.4
Conjugation: LEAF
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR