Anti-CD326 (Ep-CAM) Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: G8.8]

Supplier: BioLegend
118227 118227
10759-248EA 346.03 USD
10759-248 CA10759-248
Anti-CD326 (Ep-CAM) Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: G8.8]
Antibodies
Brilliant Violet 605™ anti-mouse CD326 (Ep-CAM) [G8.8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD326 (Ep-CAM)
Clonality: Monoclonal
Clone: G8.8
Conjugation: Brilliant Violet® 605
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR