Anti-CD326 (Ep-CAM) Rat Monoclonal Antibody (Alexa Fluor® 594) [clone: G8.8]

Supplier: BioLegend
118222 118222
10759-242EA 310.89 USD
CA10759-242 10759-242
Anti-CD326 (Ep-CAM) Rat Monoclonal Antibody (Alexa Fluor® 594) [clone: G8.8]
Antibodies
Alexa Fluor® 594 anti-mouse CD326 (Ep-CAM) [G8.8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: IF, IHC-F; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescence staining. For immunohistochemisty, a concentration range of 1-5 μg/ml is suggested. For immunofluorescence microscopy, a concentration range of 1-5 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD326 (Ep-CAM)
Clonality: Monoclonal
Clone: G8.8
Conjugation: Alexa Fluor® 594
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR