Anti-CD326 (Ep-CAM) Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: 9C4]

Supplier: BioLegend
324228 324228
10758-298EA 335.56 USD
CA10758-298 10758-298
Anti-CD326 (Ep-CAM) Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: 9C4]
Antibodies
Alexa Fluor® 594 anti-human CD326 (Ep-CAM) [9C4]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: IF, IHC-P; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescence staining. For immunofluorescence microscopy, a concentration range of 2.5-10 μg per ml is recommended. For immunohistochemical staining on formalin-fixed paraffin-embedded tissue sections, the suggested use of this reagent is 5-10 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD326 (Ep-CAM)
Clonality: Monoclonal
Clone: 9C4
Conjugation: Alexa Fluor® 594
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR