Anti-CD324 (E-Cadherin) Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: 67A4]

Supplier: BioLegend
324118 324118
10758-286EA 222.3 USD
CA10758-286 10758-286
Anti-CD324 (E-Cadherin) Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: 67A4]
Antibodies
Alexa Fluor® 594 anti-human CD324 (E-Cadherin) [67A4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: IF, IHC-P; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescence staining. For immunofluorescence microscopy, a concentration range of 2.5-10 μg/ml is recommended. For immunohistochemistry, a concentration range of 2.5 - 5.0 μg/ml is suggested. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD324 (E-Cadherin)
Clonality: Monoclonal
Clone: 67A4
Conjugation: Alexa Fluor® 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR