Anti-CD324 (E-Cadherin) Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 67A4]

Supplier: BioLegend
324116 324115 324116 324115
10758-282EA 197.13 USD
CA10758-282 10758-282 CA10758-284 10758-284
Anti-CD324 (E-Cadherin) Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 67A4]
Antibodies
PE/Cy7 anti-human CD324 (E-Cadherin) [67A4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD324 (E-Cadherin)
Clonality: Monoclonal
Clone: 67A4
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR