Anti-CD323 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SHM33]

Supplier: BioLegend
356705 356706 356705
10760-184EA 153.2 USD
10760-184 CA10760-184 CA10760-186
Anti-CD323 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SHM33]
Antibodies
APC anti-human CD323 (JAM3) [SHM33]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD323
Clonality: Monoclonal
Clone: SHM33
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR