Anti-GATA6 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP3766A
89512-302EA 637.11 USD
89512-302
Anti-GATA6 Rabbit Polyclonal Antibody
Antibodies
Immunofluorescence: 1:100

Type: Primary
Antigen: GATA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR