Anti-CD323 Mouse Monoclonal Antibody [clone: SHM33]

Supplier: BioLegend
356701 356702 356701
10760-176EA 120.94 USD
CA10760-178 10760-176 CA10760-176
Anti-CD323 Mouse Monoclonal Antibody [clone: SHM33]
Antibodies
Purified anti-human CD323 (JAM3) [SHM33]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD323
Clonality: Monoclonal
Clone: SHM33
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR