Anti-CD319 (CRACC) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 162.1]

Supplier: BioLegend
331809 331810 331809 331810
10758-466EA 160.83 USD
CA10758-466 10758-466 CA10758-468 10758-468
Anti-CD319 (CRACC) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 162.1]
Antibodies
APC anti-human CD319 (CRACC) [162.1]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cynomolgus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD319 (CRACC)
Clonality: Monoclonal
Clone: 162.1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR