Anti-CD31 Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: WM59]

Supplier: BioLegend
303126 303126
10757-770EA 360.42 USD
10757-770 CA10757-770
Anti-CD31 Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: WM59]
Antibodies
Alexa Fluor® 594 anti-human CD31 [WM59]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: IF; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescence staining. For immunofluorescence microscopy, a concentration range of 5-10 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: WM59
Conjugation: Alexa Fluor® 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR