Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDAC2 Mouse Monoclonal Antibody [clone: 1194CT18.5.1]

Supplier: Abgent

AM2246B
10217-874EA 381.69 USD InStock
10217-874
Anti-HDAC2 Mouse Monoclonal Antibody [clone: 1194CT18.5.1]
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: HDAC2
Clonality: Monoclonal
Clone: 1194CT18.5.1
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty