Anti-CD304 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: 12C2]

Supplier: BioLegend
354515 354515
10759-760EA 220.33 USD
CA10759-760 10759-760
Anti-CD304 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: 12C2]
Antibodies
Brilliant Violet 510™ anti-human CD304 (Neuropilin-1) [12C2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD304
Clonality: Monoclonal
Clone: 12C2
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR