Anti-CD304 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 12C2]

354508 354508 354507 354507
10759-746EA 147.52 USD
CA10759-746 10759-746 10759-748 CA10759-748
Anti-CD304 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 12C2]
PE/Cy7 anti-human CD304 (Neuropilin-1) [12C2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD304
Clonality: Monoclonal
Clone: 12C2
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR