Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TSC1 Mouse Monoclonal Antibody [clone: 357CT4.4.2]

Supplier: Abgent

AM1977A
89512-232EA 418.66 USD InStock
89512-232
Anti-TSC1 Mouse Monoclonal Antibody [clone: 357CT4.4.2]
Antibodies
Western Blot: 1:500~8000,

Type: Primary
Antigen: TSC1
Clonality: Monoclonal
Clone: 357CT4.4.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty