Anti-CD303 Mouse Monoclonal Antibody [clone: 201A]

Supplier: BioLegend
354202 354202
354202-BLEA 247.16 USD
CA354202-BL 354202-BL
Anti-CD303 Mouse Monoclonal Antibody [clone: 201A]
Antibodies
Purified anti-human CD303 (BDCA-2) [201A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD303
Clonality: Monoclonal
Clone: 201A
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR