Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD301b Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: URA-1]

Supplier: Biolegend

146806 146805 146805 146806
10756-562 CA10756-560 10756-560 CA10756-562
Anti-CD301b Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: URA-1]
Alexa Fluor® 647 anti-mouse CD301b (MGL2) [URA-1]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD301b
Clonality: Monoclonal
Clone: URA-1
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG2a, λ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty