Anti-CD301b Rat Monoclonal Antibody [clone: URA-1]

Supplier: BioLegend
146802 146802
10756-554EA 289.07 USD
CA10756-554 10756-554
Anti-CD301b Rat Monoclonal Antibody [clone: URA-1]
Antibodies
Purified anti-mouse CD301b (MGL2) [URA-1]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD301b
Clonality: Monoclonal
Clone: URA-1
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, λ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR