Anti-SHHA Rabbit Polyclonal Antibody

Supplier: Abcepta
AZB10041A
10217-876EA 580.86 USD
10217-876
Anti-SHHA Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: SHHA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR