Anti-CD301 Rat Monoclonal Antibody [clone: LOM-14]

Supplier: BioLegend
145702 145702
10757-450EA 278.53 USD
CA10757-450 10757-450
Anti-CD301 Rat Monoclonal Antibody [clone: LOM-14]
Antibodies
Purified anti-mouse CD301 (MGL1/MGL2) [LOM-14]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC, IP; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD301
Clonality: Monoclonal
Clone: LOM-14
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR