Anti-CD301 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: H037G3]

Supplier: BioLegend
354703 354703 354704 354704
354703-BLEA 165.11 USD
354703-BL 354704-BL CA354703-BL CA354704-BL
Anti-CD301 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: H037G3]
Antibodies
PE anti-human CD301 (CLEC10A) [H037G3]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD301
Clonality: Monoclonal
Clone: H037G3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;African Green;Baboon;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR