Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD301 Mouse Monoclonal Antibody [clone: H037G3]

Supplier: Biolegend

354702 354701 354701 354702
354702-BLEA 196.6 USD InStock
354702-BL 354701-BL CA354701-BL CA354702-BL
Anti-CD301 Mouse Monoclonal Antibody [clone: H037G3]
Antibodies
Purified anti-human CD301 (CLEC10A) [H037G3]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD301
Clonality: Monoclonal
Clone: H037G3
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;African Green;Baboon;Cynomolgus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty