Anti-CD300LG Rat Monoclonal Antibody [clone: ZAQ5]

Supplier: BioLegend
147102 147102
10757-468EA 309.51 USD
CA10757-468 10757-468
Anti-CD300LG Rat Monoclonal Antibody [clone: ZAQ5]
Antibodies
Purified anti-mouse CD300LG (Nepmucin) [ZAQ5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, EM; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD300LG
Clonality: Monoclonal
Clone: ZAQ5
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR