Anti-CD300a Rat Monoclonal Antibody [clone: TX41]

Supplier: BioLegend
147901 147901
10757-478EA 314.78 USD
CA10757-478 10757-478
Anti-CD300a Rat Monoclonal Antibody [clone: TX41]
Antibodies
Ultra-LEAF™ Purified anti-mouse CD300a [TX41]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IP, Activ; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD300a
Clonality: Monoclonal
Clone: TX41
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR