Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1F4]

Supplier: Biolegend

201413 201413 201414 201414
10756-816EA 71.03 USD InStock
10756-816 CA10756-816 10756-818 CA10756-818
Anti-CD3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1F4]
Antibodies
APC anti-rat CD3 [1F4]; Isotype: Mouse IgM, κ; Reactivity: Rat; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 1F4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgM, κ
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty