Anti-HOXD11A Rabbit Polyclonal Antibody

Supplier: Abcepta
AZB10017A
89513-502EA 636.94 USD
89513-502
Anti-HOXD11A Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: HOXD11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR