Anti-CD3 Rat Monoclonal Antibody (Brilliant Violet® 650) [clone: 17A2]

Supplier: BioLegend
100229 100229
100229-BLEA 315.9 USD
100229-BL CA100229-BL
Anti-CD3 Rat Monoclonal Antibody (Brilliant Violet® 650) [clone: 17A2]
Antibodies
Brilliant Violet 650™ anti-mouse CD3 [17A2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 125 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 17A2
Conjugation: Brilliant Violet® 650
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR