Anti-CD3 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: SK7]

Supplier: BioLegend
344825 344826 344826 344825
10758-742EA 342.97 USD
10758-742 CA10758-742 10758-740 CA10758-740
Anti-CD3 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: SK7]
Antibodies
Alexa Fluor® 647 anti-human CD3 [SK7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: SK7
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR