Anti-CD3 Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: OKT3]

Supplier: BioLegend
317341 317342 317342 317341
10758-198EA 465.89 USD
CA10758-198 CA10758-196 10758-198 10758-196
Anti-CD3 Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: OKT3]
Antibodies
APC/Cy7 anti-human CD3 [OKT3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: OKT3
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR