Anti-CD294 Rat Monoclonal Antibody [clone: BM16]

Supplier: BioLegend
350102 350102
350102-BLEA 135.97 USD
CA350102-BL 350102-BL
Anti-CD294 Rat Monoclonal Antibody [clone: BM16]
Antibodies
Purified anti-human CD294 (CRTH2) [BM16]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD294
Clonality: Monoclonal
Clone: BM16
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR