Anti-CD29 Mouse Monoclonal Antibody [clone: TS2/16]

Supplier: BioLegend
303021 303021
10759-456EA 157.98 USD
CA10759-456 10759-456
Anti-CD29 Mouse Monoclonal Antibody [clone: TS2/16]
Antibodies
Purified anti-human CD29 (MaxPar® Ready) [TS2/16]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cattle (Bovine, Cow), Cynomolgus, Dog (Canine), (Horse reactivity), Rhesus; Apps: FC, CyTOF®; Size: 100 μg

This product is suitable for use with the <a href="http://www.dvssciences.com/labeling-reagents.php" target="_blank">MaxPar® Metal Labeling Kits</a>. The product is formulated so that the buffer exchange step is not required (steps 7, 8, 9, and 11 in the <a href="http://www.dvssciences.com/technical-literature_13_3283624161.pdf" target="_blank">MaxPar® antibody labeling protocol</a>). Just add 100 μl of this antibody to 100 μl of 4 mM TCEP-R in a 50 kDa filter, as described in step 12, and continue with the protocol.

Type: Primary
Antigen: CD29
Clonality: Monoclonal
Clone: TS2/16
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cattle (Bovine;Cow);Cynomolgus;Dog (Canine);(Horse reactivity);Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR