Anti-CD284 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: SA15-21]

Supplier: BioLegend
145404 145403 145404 145403
10756-468EA 131.66 USD
CA10756-468 10756-468 CA10756-470 10756-470
Anti-CD284 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: SA15-21]
Antibodies
PE anti-mouse CD284 (TLR4) [SA15-21]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD284
Clonality: Monoclonal
Clone: SA15-21
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR