Anti-CD282 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: CB225]

Supplier: BioLegend
148604 148603 148604 148603
10756-794EA 374.62 USD
CA10756-794 10756-794 CA10756-792 10756-792
Anti-CD282 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: CB225]
Antibodies
PE anti-mouse CD282 (TLR2) [CB225]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD282
Clonality: Monoclonal
Clone: CB225
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR