Anti-CD28 Syrian Hamster Monoclonal Antibody [clone: 37.51]

Supplier: BioLegend
102119 102119
10756-780EA 114.47 USD
CA10756-780 10756-780
Anti-CD28 Syrian Hamster Monoclonal Antibody [clone: 37.51]
Antibodies
Purified anti-mouse CD28 (MaxPar® Ready) [37.51]; Isotype: Syrian Hamster IgG; Reactivity: Mouse; Apps: FC, CyTOF®; Size: 100 μg

This product is suitable for use with the <a href="http://www.dvssciences.com/labeling-reagents.php" target="_blank">MaxPar® Metal Labeling Kits</a>. The product is formulated so that the buffer exchange step is not required (steps 7, 8, 9, and 11 in the <a href="http://www.dvssciences.com/technical-literature_13_3283624161.pdf" target="_blank">MaxPar® antibody labeling protocol</a>). Just add 100 μl of this antibody to 100 μl of 4 mM TCEP-R in a 50 kDa filter, as described in step 12, and continue with the protocol.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: 37.51
Conjugation:
Epitope:
Host: Syrian Hamster
Isotype: Syrian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR