Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BLNK Mouse Monoclonal Antibody [clone: 520CT6.1.1]

Supplier: Abgent

AM2071A
89512-142
Anti-BLNK Mouse Monoclonal Antibody [clone: 520CT6.1.1]
Western Blot: 1:500~8000,

Type: Primary
Antigen: BLNK
Clonality: Monoclonal
Clone: 520CT6.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty