Anti-CD279 (PD-1) Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: EH12.2H7]

Supplier: BioLegend
329928 329927 329928 329927
10758-404EA 611.37 USD
10758-404 CA329927-BL CA10758-404 329927-BL
Anti-CD279 (PD-1) Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: EH12.2H7]
Antibodies
Brilliant Violet 711™ anti-human CD279 (PD-1) [EH12.2H7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD279 (PD-1)
Clonality: Monoclonal
Clone: EH12.2H7
Conjugation: Brilliant Violet® 711
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Chimpanzee;Common Marmoset;Cynomolgus;Rhesus;Squirrel Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR