Anti-RAB2B Rabbit Polyclonal Antibody

Supplier: Abcepta
AP17933C
89517-402EA 636.61 USD
89517-402
Anti-RAB2B Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: RAB2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR