Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD278 (ICOS) Armenian Hamster Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: C398.4A]

Supplier: Biolegend

313517 313517 313518 313518
313517-BL CA313517-BL 313518-BL CA313518-BL
Anti-CD278 (ICOS) Armenian Hamster Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: C398.4A]
PerCP/Cy5.5 anti-human/mouse/rat CD278 (ICOS) [C398.4A]; Isotype: Armenian Hamster IgG; Reactivity: Human, African Green, Baboon, Cynomolgus, Mouse, Rat, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application. <br />* PerCP/Cy5.5 has a maximum absorption of 482 nm and a maximum emission of 690 nm.

Type: Primary
Antigen: CD278 (ICOS)
Clonality: Monoclonal
Clone: C398.4A
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Human;African Green;Baboon;Cynomolgus;Mouse;Rat;Rhesus;Swine (Pig;Porcine)

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty