Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SLC16A10 Rabbit Polyclonal Antibody

Supplier: Abgent

AP13235B
89512-980EA 418.66 USD InStock
89512-980
Anti-SLC16A10 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:2000, Immunhistochemsitry: 1:10~50, Flow Cytometry: 1:10~50,

Type: Primary
Antigen: SLC16A10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty