Anti-CREB3L4 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP16716B
89519-436EA 637.11 USD
89519-436
Anti-CREB3L4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CREB3L4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR