Anti-CD27 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: M-T271]

Supplier: BioLegend
356405 356405 356406 356406
10759-976EA 83.15 USD
10759-976 10759-978 CA10759-978 CA10759-976
Anti-CD27 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: M-T271]
Antibodies
PE anti-human CD27 [M-T271]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: M-T271
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR