Anti-CD25 Mouse Monoclonal Antibody (Biotin) [clone: M-A251]

Supplier: BioLegend
356123 356124 356124 356123
10759-930EA 222.3 USD
CA10759-928 10759-930 CA10759-930 10759-928
Anti-CD25 Mouse Monoclonal Antibody (Biotin) [clone: M-A251]
Antibodies
Biotin anti-human CD25 [M-A251]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: M-A251
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR