Anti-CD25 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: M-A251]

Supplier: BioLegend
356107 356108 356107 356108
10759-896EA 174.9 USD
CA10759-896 10759-896 10759-898 CA10759-898
Anti-CD25 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: M-A251]
Antibodies
PE/Cy7 anti-human CD25 [M-A251]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: M-A251
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR