Anti-CD243 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: UIC2]

Supplier: BioLegend
348609 348610 348610
10759-566EA 374.65 USD
CA10759-566 10759-566 CA10759-388
Anti-CD243 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: UIC2]
Antibodies
PE/Cy7 anti-human CD243 (MDR-1) [UIC2]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD243
Clonality: Monoclonal
Clone: UIC2
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;African Green;Baboon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR