Anti-CD235ab Mouse Monoclonal Antibody [clone: HIR2]

Supplier: BioLegend
306615 306615
10757-956EA 139.32 USD
10757-956 CA10757-956
Anti-CD235ab Mouse Monoclonal Antibody [clone: HIR2]
Antibodies
Purified anti-human CD235ab (MaxPar® Ready) [HIR2]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon; Apps: FC, CyTOF®; Size: 100 μg

This product is suitable for use with the <a href="http://www.dvssciences.com/labeling-reagents.php" target="_blank">MaxPar® Metal Labeling Kits</a>. The product is formulated so that the buffer exchange step is not required (steps 7, 8, 9, and 11 in the <a href="http://www.dvssciences.com/technical-literature_13_3283624161.pdf" target="_blank">MaxPar® antibody labeling protocol</a>). Just add 100 μl of this antibody to 100 μl of 4 mM TCEP-R in a 50 kDa filter, as described in step 12, and continue with the protocol.

Type: Primary
Antigen: CD235ab
Clonality: Monoclonal
Clone: HIR2
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;African Green;Baboon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR