Anti-PIP4K2C Rabbit Polyclonal Antibody

Supplier: Abcepta
AP20635A
10218-260EA 580.54 USD
10218-260
Anti-PIP4K2C Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: PIP4K2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR