Anti-CD235a Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HI264]

Supplier: BioLegend
349106 349105 349105 349106
349105-BLEA 169.47 USD
349105-BL CA349106-BL 349106-BL CA349105-BL
Anti-CD235a Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HI264]
Antibodies
PE anti-human CD235a (Glycophorin A) [HI264]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD235a
Clonality: Monoclonal
Clone: HI264
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR